Phenylmagnesium bromide 1.6 M in CPME (cyclopentyl methyl ether), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC44596-1000 AC44596-8000
TS44596-1000EA 119 USD
TS44596-1000 TS44596-8000
Phenylmagnesium bromide 1.6 M in CPME (cyclopentyl methyl ether), AcroSeal™
Phenylmagnesium bromide
Formula: C₆H₅BrMg
MW: 181.31 g/mol
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000038
CAS Number: 100-58-3
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to orange Solution (May develop some turbidity or precipitate)
Titration Grignard 1.50 to 1.70 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR