Hexachloroiridic(IV) acid hexahydrate (∼40% Ir)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexachloroiridate(IV) hexahydrate
AC19755-0010
AC19755-0050
TS19755-0010EA
327
USD
TS19755-0010
TS19755-0050
Hexachloroiridic(IV) acid hexahydrate (∼40% Ir)
Hexachloroiridic(IV) acid hexahydrate
Formula:
H₂Cl₆Ir·6H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149907 CAS Number: 16941-92-7 EINECS: 241-012-8 UN: 3260 ADR: 8 III |
Specification Test Results
Appearance | Black Powder or crystals |
Assay | 37 to 43 % (Ir) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...