4-(2-Chloroethyl)morpholine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride
AC10920-0050
AC10920-1000
AC10920-5000
TS10920-0050EA
30
USD
TS10920-0050
TS10920-1000
TS10920-5000
4-(2-Chloroethyl)morpholine hydrochloride 99%
4-(2-Chloroethyl)morpholine hydrochloride
Formula:
C₆H₁₃Cl₂NO MW: 186.08 g/mol Melting Pt: 182…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012797 CAS Number: 3647-69-6 EINECS: 222-881-2 UN: 2923 ADR: 8,III |
Specification Test Results
Appearance | White to beige Hygroscopic crystalline powder |
Infrared spectrum | Conforms |
Melting point | 182 to 185 °C (on dry substance) |
Loss on drying | ≤1 % (80 °C) |
Titration with HClO4 | ≥98.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...