Mercury (II) oxide, yellow ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC31679-1000
AC31679-5000
TS31679-1000EA
109
USD
TS31679-1000
TS31679-5000
Mercury (II) oxide, yellow ACS
Mercury (II) oxide
Formula:
HgO Melting Pt: 500 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011045 CAS Number: 21908-53-2 EINECS: 244-654-7 UN: 1641 ADR: 6.1,II Merck Index: 13,05908 |
Specification Test Results
Appearance | Yellow to light orange Powder |
Titration Complexometric | ≥99.0 % |
Chloride (Cl) | ≤250 ppm |
Sulfate (SO4) | ≤100 ppm |
Iron (Fe) | ≤30 ppm |
Insoluble matter | ≤0.03 % (in dilute HCl) |
Residue after reduction | ≤0.05 % |
Nitrogen compounds (as N) | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...