Ethylmagnesium bromide 40% 3 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC34729-1000 AC34729-8000
TS34729-1000EA 73 USD
TS34729-1000 TS34729-8000
Ethylmagnesium bromide 40% 3 M in diethyl ether, AcroSeal™
Ethylmagnesium bromide
Formula: C₂H₅BrMg
MW: 133.27 g/mol
Density: 1.03 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Dark brown Solution
Titration Grignard 2.80 to 3.20 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR