Ethylmagnesium bromide ∼3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC18353-1000 AC18353-8000
TS18353-1000EA 104 USD
TS18353-1000 TS18353-8000
Ethylmagnesium bromide ∼3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Ethylmagnesium bromide
Formula: C₂H₅BrMg
MW: 133.27 g/mol
Density: 1.14 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 39 to 41 %
Water ≤1 %
Solubility (2 % in DMSO) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR