Ethylmagnesium bromide 0.9 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC21047-1000 AC21047-8000
TS21047-1000EA 65 USD
TS21047-1000 TS21047-8000
Ethylmagnesium bromide 0.9 M in THF, AcroSeal™
Ethylmagnesium bromide
Formula: C₂H₅BrMg
MW: 133.27 g/mol
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Brown to gray-brown to brown-black Solution (May develop some turbidity or precipitate)
Titration Grignard 0.85 to 1.15 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR