Sodium tetrachloroaurate hydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC39627-0010
AC39627-0050
TS39627-0010EA
149
USD
TS39627-0010
TS39627-0050
Sodium tetrachloroaurate hydrate 98%
Sodium tetrachloroaurate hydrate
Formula:
AuCl₄Na·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149161 CAS Number: 29156-65-8 UN: 1759 ADR: 8,III |
Specification Test Results
Appearance | Yellow to orange to golden Powder or crystals or granules |
Assay | 48.3 to 50.3 % (Au) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...