Ethylaluminium dichloride 1.8 M in toluene, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42804-1000 AC42804-8000
TS42804-1000EA 86 USD
TS42804-1000 TS42804-8000
Ethylaluminium dichloride 1.8 M in toluene, AcroSeal™
Ethylaluminium dichloride
Formula: C₂H₅AlCl₂
MW: 126.95 g/mol
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000457
CAS Number: 563-43-9
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Strength 23 to 27 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR