Ethylaluminium dichloride 0.9 M in heptane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC18545-1000 AC18545-8000
TS18545-1000EA 87 USD
TS18545-1000 TS18545-8000
Ethylaluminium dichloride 0.9 M in heptane, AcroSeal™
Ethylaluminium dichloride
Formula: C₂H₅AlCl₂
Density: 0.74 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000457
CAS Number: 563-43-9
EINECS: 209-248-6
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Assay 14.5 to 15.5 %
Loss on drying ≤4 %
HPLC ≥97.5 % (mixture of A and B)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR