Cacodylic acid (dimethylarsinic acid) 98%

Supplier: Thermo Scientific Chemicals
Danger

AC31815-0050 AC31815-0100 AC31815-0500 AC31815-2500
TS31815-0050EA 57 USD
TS31815-0050 TS31815-0100 TS31815-0500 TS31815-2500
Cacodylic acid (dimethylarsinic acid) 98%
Cacodylic acid (dimethylarsinic acid)
Formula: (CH₃)₂As(O)OH
Melting Pt: 195…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00002095
CAS Number: 75-60-5
EINECS: 200-883-4
UN: 1572
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 195 to 198 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR