Dichloro(chloromethyl)methylsilane 95%

Supplier: Thermo Scientific Chemicals
Danger

AC31337-0250 AC31337-1000
TS31337-0250EA 48 USD
TS31337-0250 TS31337-1000
Dichloro(chloromethyl)methylsilane 95%
Dichloro(chloromethyl)methylsilane
Formula: C₂H₅Cl₃Si
MW: 163.51 g/mol
Boiling Pt: 121…122 °C
Density: 1.26 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00000874
CAS Number: 1558-33-4
EINECS: 216-319-5
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR