Allyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

AC22174-0500 AC22174-1000
TS22174-0500EA 135 USD
TS22174-0500 TS22174-1000
Allyl chloroformate 97%
Allyl chloroformate
Formula: C₄H₅ClO₂
Boiling Pt: 109…110 °C (760 mmHg)
Density: 1.13 g/cm³
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000648
CAS Number: 2937-50-0
EINECS: 220-916-6
UN: 1722
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4240 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR