Ethyl isobutyrylacetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-methyl-3-oxopentanoate

AC38344-0100 AC38344-0500
TS38344-0100EA 104 USD
TS38344-0100 TS38344-0500
Ethyl isobutyrylacetate 95%
Ethyl isobutyrylacetate
Formula: C₈H₁₄O₃
Boiling Pt: 173 °C (760 mmHg)
Melting Pt: –9 °C
Density: 0.981 g/cm³
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00009198
CAS Number: 7152-15-0
EINECS: 230-491-9
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4280 (20°C, 589 nm)
GC ≥94 % (Mixture of tautomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR