Trimethyl citrate
Supplier: Thermo Scientific Chemicals
Synonyms:
Citric acid trimethyl ester
AC46225-0050
AC46225-0250
TS46225-0050EA
39
USD
TS46225-0050
TS46225-0250
Trimethyl citrate
Trimethyl citrate
Formula:
C₉H₁₄O₇ MW: 234.21 g/mol Boiling Pt: 176 °C (16 mmHg) Melting Pt: 75…78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00025889 CAS Number: 1587-20-8 |
Specification Test Results
Appearance | White to off-white Crystals |
Infrared spectrum | Conforms |
Melting point | 75 to 82 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...