Potassium thioacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium ethanethioate

AC22130-0250 AC22130-1000 AC22130-5000
TS22130-0250EA 64 USD
TS22130-0250 TS22130-1000 TS22130-5000
Potassium thioacetate 98%
Potassium thioacetate
Formula: C₂H₃KOS
MW: 114.21 g/mol
Melting Pt: 173…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137704
CAS Number: 10387-40-3
EINECS: 233-848-7
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR