Aluminium tri-sec-butanolate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

AC19121-0025 AC19121-1000 AC19121-5000
TS19121-0025EA 256 USD
TS19121-0025 TS19121-1000 TS19121-5000
Aluminium tri-sec-butanolate 97%
Aluminium tri-sec-butanolate
Formula: C₁₂H₂₇AlO₃
MW: 246.33 g/mol
Boiling Pt: 200…206 °C (30 mmHg)
Density: 0.96 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00009327
CAS Number: 2269-22-9
EINECS: 218-871-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
Assay ≥96.0 % (ex Al)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR