Bis(cyclopentadienyl)zirconium hydridochloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride

AC20789-0010 AC20789-0050 AC20789-0250
TS20789-0010EA 42 USD
TS20789-0010 TS20789-0050 TS20789-0250
Bis(cyclopentadienyl)zirconium hydridochloride 95%
Bis(cyclopentadienyl)zirconium hydridochloride
Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02089401
CAS Number: 37342-97-5
EINECS: 253-479-5
UN: 3131
ADR: 4.3,II

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration Argentometric ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR