Sodium benzenesulfinate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzenesulphinic acid sodium salt, Benzenesulphinic acid, sodium salt
AC15405-1000
AC15405-5000
TS15405-1000EA
73
USD
TS15405-1000
TS15405-5000
Sodium benzenesulfinate 97%
Sodium benzenesulphinate
Formula:
C₆H₅NaO₂S MW: 164.16 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013135 CAS Number: 873-55-2 EINECS: 212-842-8 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | 96.0 to 104.0 % |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...