Cysteaminium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl
AC15377-0050
AC15377-0250
AC15377-1000
AC15377-5000
TS15377-0050EA
30
USD
TS15377-0050
TS15377-0250
TS15377-1000
TS15377-5000
Cysteaminium chloride 98%
Cysteaminium chloride
Formula:
C₂H₇NS·HCl Melting Pt: 66…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012904 CAS Number: 156-57-0 EINECS: 205-858-1 UN: 3335 ADR: 9,III |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or lumps |
Infrared spectrum | Conforms |
Melting point | 66 to 70 °C |
Loss on drying | ≤1 % (50 °C, 4 h, 100 mmHg) |
Titration with KIO3 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...