1-(2-Chloroethyl)piperidinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

AC10921-1000 AC10921-5000
TS10921-1000EA 46 USD
TS10921-1000 TS10921-5000
1-(2-Chloroethyl)piperidinium chloride 98%
1-(2-Chloroethyl)piperidinium chloride
Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
Melting Pt: 228…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00012837
CAS Number: 2008-75-5
EINECS: 217-920-5
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 228 to 234 °C
Loss on drying ≤1 % (105 °C, 3 hrs)
Additional info total chloride: 38.0 to 39.0 % w/w
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR