(Triisopropylsilyl)acetylene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethynyltriisopropylsilane

AC36874-0050 AC36874-0250
TS36874-0050EA 85 USD
TS36874-0050 TS36874-0250
(Triisopropylsilyl)acetylene 97%
(Triisopropylsilyl)acetylene
Formula: C₁₁H₂₂Si
MW: 182.38 g/mol
Boiling Pt: 50…52 °C (6 mmHg)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00075452
CAS Number: 89343-06-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4515 to 1.4535 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR