(tert-Butyldimethylsilyl)acetylene 97%

Supplier: Thermo Scientific Chemicals
Danger

AC38130-0010 AC38130-0050
TS38130-0010EA 72 USD
TS38130-0010 TS38130-0050
(tert-Butyldimethylsilyl)acetylene 97%
(tert-Butyldimethylsilyl)acetylene
Formula: C₈H₁₆Si
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00191877
CAS Number: 86318-61-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR