Benzyl chloroformate 97% (w/w) stabilized

Supplier: Thermo Scientific Chemicals
Danger

AC15294-0025 AC15294-0050 AC15294-1000 AC15294-5000
TS15294-0025EA 725 USD
TS15294-0025 TS15294-0050 TS15294-1000 TS15294-5000
Benzyl chloroformate 97% (w/w) stabilized
Benzyl chloroformate
Formula: C₈H₇ClO₂
Boiling Pt: 178…180 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.2 g/cm³
Flash Pt: 92 °C
Storage Temperature: Freezer
MDL Number: MFCD00000640
CAS Number: 501-53-1
EINECS: 207-925-0
UN: 1739
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
Additional info benzyl chloride during storage
GC ≥96.0 %
Color scale ≤100 APHA
Stabilizer 0.1 % anhydrous sodium carbonate
Phosgene ≤0.1 %
Iron (Fe) ≤2 ppm
Acidity (HCl) ≤0.3 %
Benzyl alcohol ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR