Magnesium ethanolate 98%

Supplier: Thermo Scientific Chemicals
Danger

AC39747-0050 AC39747-1000 AC39747-5000
TS39747-0050EA 26 USD
TS39747-0050 TS39747-1000 TS39747-5000
Magnesium ethanolate 98%
Magnesium ethanolate
Formula: C₄H₁₀MgO₂
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009060
CAS Number: 2414-98-4
UN: 3205
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to light gray Powder
Infrared spectrum Conforms
Additional info May contain up to 2.5% Mg(OH)2 and/or MgCO3
Titration with HCl 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR