Zinc trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate
AC31709-0100
AC31709-0500
TS31709-0100EA
59
USD
TS31709-0100
TS31709-0500
Zinc trifluoromethanesulfonate 98%
Zinc trifluoromethanesulphonate
Formula:
C₂F₆O₆S₂Zn MW: 363.53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013229 CAS Number: 54010-75-2 EINECS: 258-922-6 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White to light gray Powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...