Ytterbium (III) trifluoromethanesulfonate, anhydrous 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ytterbium (III) triflate
AC43405-0010
AC43405-0050
TS43405-0010EA
39
USD
TS43405-0010
TS43405-0050
Ytterbium (III) trifluoromethanesulfonate, anhydrous 99.9% (trace metals basis)
Ytterbium(III) trifluoromethanesulphonate
Formula:
C₃F₉O₉S₃Yb MW: 620.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06200261 CAS Number: 54761-04-5 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration Complexometric | 26.8 to 29.0 % (Yb) |
Total conc. of imp.as REO | ≤1000 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...