Samarium (III) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Samarium(III) trifluoromethanesulphonate

AC38317-0010 AC38317-0100
TS38317-0010EA 37 USD
TS38317-0010 TS38317-0100
Samarium (III) trifluoromethanesulfonate 98%
Samarium(III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192431
CAS Number: 52093-28-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis 24.65 to 25.66 % (Sm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR