Lanthanum(III) trifluoromethanesulphonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate
AC34391-0010
AC34391-0050
TS34391-0010EA
37
USD
TS34391-0010
TS34391-0050
Lanthanum(III) trifluoromethanesulphonate 99%
Lanthanum(III) trifluoromethanesulphonate
Formula:
C₃F₉LaO₉S₃ MW: 586.12 g/mol Density: 1.7 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00077560 CAS Number: 52093-26-2 UN: 3261 ADR: 8 II |
Specification Test Results
Appearance | White to light beige Powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | 23.35 to 24.05 % (La) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...