Tin(II) trifluoromethanesulphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

AC31253-0010 AC31253-0050
TS31253-0010EA 66 USD
TS31253-0010 TS31253-0050
Tin(II) trifluoromethanesulphonate 98%
Tin(II) trifluoromethanesulphonate
Formula: C₂F₆O₆S₂Sn
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191251
CAS Number: 62086-04-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Sn)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR