Bis(triphenylphosphine)nickel(II) chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

AC21750-0100 AC21750-0500
TS21750-0100EA 54 USD
TS21750-0100 TS21750-0500
Bis(triphenylphosphine)nickel(II) chloride 98%
Bis(triphenylphosphine)nickel(II) chloride
Formula: C₃₆H₃₂Cl₂NiP₂
MW: 656.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00009592
CAS Number: 14264-16-5
EINECS: 238-154-8
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance Dark green to dark gray Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR