Tris(triphenylphosphine)ruthenium(II) chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)
AC20788-0010
AC20788-0050
AC20788-0250
TS20788-0010EA
55
USD
TS20788-0010
TS20788-0050
TS20788-0250
Tris(triphenylphosphine)ruthenium(II) chloride 98%
Tris(triphenylphosphine)ruthenium(II) chloride
Formula:
C₅₄H₄₅Cl₂P₃Ru MW: 958.85 g/mol Melting Pt: 159 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013077 CAS Number: 15529-49-4 EINECS: 239-569-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Red-brown to black Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Carbon (C) | 66.0 to 69.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...