4-Picolyl bromide hydrobromide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)pyridine hydrobromide

AC39733-0050 AC39733-0250
TS39733-0050EA 60 USD
TS39733-0050 TS39733-0250
4-Picolyl bromide hydrobromide 97%
4-Picolyl bromide hydrobromide
Formula: C₆H₇Br₂N
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD01863545
CAS Number: 73870-24-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 189 to 192 °C
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR