2-Picolyl bromide hydrobromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Bromomethyl)pyridine hydrobromide

AC37011-0050 AC37011-0250 AC37011-1000
TS37011-0050EA 49 USD
TS37011-0050 TS37011-0250 TS37011-1000
2-Picolyl bromide hydrobromide 98%
2-Picolyl bromide hydrobromide
Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD01863544
CAS Number: 31106-82-8
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White to pink to orange-brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR