4-Chloropyridine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

AC11003-0250 AC11003-1000
TS11003-0250EA 87 USD
TS11003-0250 TS11003-1000
4-Chloropyridine hydrochloride 97%
4-Chloropyridine hydrochloride
Formula: C₅H₅Cl₂N
Melting Pt: 210 °C (subl.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00012829
CAS Number: 7379-35-3
EINECS: 230-946-1

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR