Selenourea 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC21317-0010 AC21317-0050
TS21317-0010EA 79 USD
TS21317-0010 TS21317-0050
Selenourea 98+%
Selenourea
Formula: CH₄N₂Se
Melting Pt: 214…215 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00008065
CAS Number: 630-10-4
EINECS: 211-129-9
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to pink to gray Powder or crystals or needles
Infrared spectrum Conforms
Assay ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR