Dichloro(diphenyl)silane 97%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diphenyldichlorosilane, Dichlorodiphenylsilane

AC43089-1000 AC43089-8000
TS43089-1000EA 48 USD
TS43089-1000 TS43089-8000
Dichloro(diphenyl)silane 97%, AcroSeal™
Dichloro(diphenyl)silane
Formula: C₁₂H₁₀Cl₂Si
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.204 g/cm³
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00000489
CAS Number: 80-10-4
UN: 1769
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5765 to 1.5815 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR