Tetramethylammonium hydrogen sulphate 99+% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetramethylammonium hydrogensulphate, Tetramethylammonium bisulphate
AC34383-0050
AC34383-0500
TS34383-0050EA
77
USD
TS34383-0050
TS34383-0500
Tetramethylammonium hydrogen sulphate 99+% for HPLC
Tetramethylammonium hydrogen sulphate
Formula:
C₄H₁₃NO₄S MW: 171.22 g/mol Melting Pt: 295…297 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036149 CAS Number: 80526-82-5 EINECS: 279-490-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
UV | (0.005 M in water) |
Titration with NaOH | ≥99.0 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...