2-(Trimethylsilyl)thiazole 96%
Supplier: Thermo Scientific Chemicals
AC29229-0050
AC29229-0250
TS29229-0050EA
184
USD
TS29229-0050
TS29229-0250
2-(Trimethylsilyl)thiazole 96%
2-(Trimethylsilyl)thiazole
Formula:
C₆H₁₁NSSi Boiling Pt: 53…55 °C (10 mmHg) Density: 0.98 g/cm³ Flash Pt: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066274 CAS Number: 79265-30-8 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4950 to 1.5000 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...