Trimethylsilyl isocyanate 94%

Supplier: Thermo Scientific Chemicals
Danger

AC22473-0050 AC22473-0250
TS22473-0050EA 97 USD
TS22473-0050 TS22473-0250
Trimethylsilyl isocyanate 94%
Trimethylsilyl isocyanate
Formula: C₄H₉NOSi
Melting Pt: –49 °C
Flash Pt: –2 °C
Storage Temperature: Freezer
MDL Number: MFCD00001993
CAS Number: 1118-02-1
EINECS: 214-256-8
UN: 2478
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR