Silver trifluoroacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silver(I) trifluoroacetate

AC13973-0050 AC13973-0250
TS13973-0050EA 67 USD
TS13973-0050 TS13973-0250
Silver trifluoroacetate 98%
Silver trifluoroacetate
Formula: C₂AgF₃O₂
MW: 220.88 g/mol
Melting Pt: 257…260 °C
MDL Number: MFCD00013199
CAS Number: 2966-50-9
EINECS: 221-004-0
UN: 3077

Order Now

Specification Test Results

Appearance Light beige to gray Crystalline powder
Infrared spectrum Conforms
Titration with NH4SCN ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR