Sodium dihydrogen citrate, anhydrous 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate

AC21533-0025 AC21533-1000 AC21533-5000
TS21533-0025EA 280 USD
TS21533-0025 TS21533-1000 TS21533-5000
Sodium dihydrogen citrate, anhydrous 99%, pure
Sodium dihydrogen citrate
Formula: C₆H₇NaO₇
MW: 214.11 g/mol
Melting Pt: 209…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS Number: 18996-35-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR