Disodium hydrogen citrate sesquihydrate 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate

AC25024-0010 AC25024-0025 AC25024-2500
TS25024-0010EA 112 USD
TS25024-0010 TS25024-0025 TS25024-2500
Disodium hydrogen citrate sesquihydrate 99%, pure
di-Sodium hydrogen citrate sesquihydrate
Formula: C₆H₆Na₂O₇·1.5H₂O
MW: 263.11 g/mol
Melting Pt: 149 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150445
CAS Number: 6132-05-4
EINECS: 242-734-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
pH 4.9 to 5.2 (3 % solution)
Titration with HClO4 98.5 to 104.0 %
Insoluble matter ≤0.01 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR