Zinc(II) acetylacetonate (∼25% Zn)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II), Zinc acetylacetonate, Zinc (II) 2,4-pentanedionate, Zinc (II) bis(2,4-pentanedionato-O,O')

AC38877-0050 AC38877-1000 AC38877-5000
TS38877-0050EA 27 USD
TS38877-0050 TS38877-1000 TS38877-5000
Zinc(II) acetylacetonate (∼25% Zn)
Zinc(II) acetylacetonate
Formula: C₁₀H₁₄O₄Zn
MW: 263.61 g/mol
Boiling Pt: 129…131 °C (10 mmHg)
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00000035
CAS Number: 14024-63-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Complexometric 23.5 to 26.0 % (Zinc)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR