Potassium cyclopropyltrifluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclopropyltrifluoroboric acid potassium salt
AC44487-0010
AC44487-0050
TS44487-0010EA
104
USD
TS44487-0010
TS44487-0050
Potassium cyclopropyltrifluoroborate 97%
Potassium cyclopropyltrifluoroborate
Formula:
C₃H₅BF₃K MW: 147.98 Melting Pt: 348…350 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09265031 CAS Number: 1065010-87-8 |
Specification Test Results
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...