Propargyltrimethylsilane 80-90% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethyl(propargyl)silane, 3-(Trimethylsilyl) propyne, Trimethyl(prop-2-yn-1-yl)silane

AC36663-0010 AC36663-0050
TS36663-0010EA 223 USD
TS36663-0010 TS36663-0050
Propargyltrimethylsilane 80-90% stabilized
Propargyltrimethylsilane
Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 91…93 °C (760 mmHg)
Density: 0.75 g/cm³
Flash Pt: –10 °C
Storage Temperature: Freezer
MDL Number: MFCD00042922
CAS Number: 13361-64-3
EINECS: 236-427-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC 80 to 90 %
Stabilizer 0.1 % hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR