1-(Trimethylsilyl)-1-propyne 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethyl-1-propynylsilane

AC22353-0010 AC22353-0050
TS22353-0010EA 36 USD
TS22353-0010 TS22353-0050
1-(Trimethylsilyl)-1-propyne 98%
1-(Trimethylsilyl)-1-propyne
Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 99…100 °C (760 mmHg)
Density: 0.75 g/cm³
Flash Pt: –3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009271
CAS Number: 6224-91-5
EINECS: 228-314-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR