Tributylphenyltin
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC37023-0010
AC37023-0100
AC37023-0500
TS37023-0010EA
39
USD
TS37023-0010
TS37023-0100
TS37023-0500
Tributylphenyltin
Tributylphenyltin
Formula:
C₁₈H₃₂Sn Boiling Pt: 125…128 °C (.1 mmHg) Density: 1.125 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134394 CAS Number: 960-16-7 UN: 2788 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5170 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...