Tributylvinyltin 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tri-n-butylvinyltin, Tributylvinylstannane
AC35000-0010
AC35000-0050
AC35000-0250
TS35000-0010EA
47
USD
TS35000-0010
TS35000-0050
TS35000-0250
Tributylvinyltin 97%
Tributylvinyltin
Formula:
C₁₄H₃₀Sn MW: 317.1 g/mol Boiling Pt: 104…106 °C (3.5 mmHg) Density: 1.08 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009421 CAS Number: 7486-35-3 EINECS: 231-291-4 UN: 2788 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4790 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...