Dipotassium hexachloroosmate > 39% Os

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium hexachloroosmate(IV)

AC19765-0010 AC19765-0050
TS19765-0010EA 187 USD
TS19765-0010 TS19765-0050
Dipotassium hexachloroosmate > 39% Os
Dipotassium hexachloroosmate
Formula: Cl₆K₂Os
MW: 481.14 g/mol
Melting Pt: 600 °C
MDL Number: MFCD00011371
CAS Number: 16871-60-6
EINECS: 240-893-6
UN: 3290

Order Now

Specification Test Results

Appearance Bordeaux to purple Powder
Assay ≥39 % (Os)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR